EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
Intraperitoneal and subcutaneous injections of the TLR9 agonist ODN 1668 in rats: brain inflammatory responses are related to peripheral IL-6 rather than interferons.
PMID 25465287
Abstract
Subcutaneous or intraperitoneal administration of Toll-like receptor (TLR)-9 agonist, ODN 1668 caused moderate fever and anorexia. In comparison to stimulation of other intracellular TLRs, activation of TLR9 did not result in pronounced peripheral induction of interferons, but rather induced interleukin-6. Expression of cytokines (TNFα, IL-1β) and inducible forms of enzymes for prostaglandin E2 synthesis occurred in the brain, in conjunction with a moderate activation of the transcription factors STAT3 and NF-IL6 in brain endothelial cells. The lack of a septic-like state in ODN 1668-treated rats reinforces the therapeutic value of this drug.