EMAIL THIS PAGE TO A FRIEND
Metabolism: clinical and experimental
Dysfunctional high-density lipoproteins in children with chronic kidney disease.
PMID 25467845
Abstract
Our aim was to determine if chronic kidney disease (CKD) occurring in childhood impairs the normally vasoprotective functions of high-density lipoproteins (HDLs). HDLs were isolated from children with end-stage renal disease on dialysis (ESRD), children with moderate CKD and controls with normal kidney function. Macrophage response to HDLs was studied as expression of inflammatory markers (MCP-1, TNF-α, IL-1β) and chemotaxis. Human umbilical vein endothelial cells were used for expression of adhesion molecules (ICAM-1, VCAM-1, E-selectin) and adhesion. Cellular proliferation, apoptosis, and necrosis of endothelial cells were measured by MTS/PMS reagent-based assay, flow cytometry, and ELISA. Cholesterol efflux was assessed by gas chromatographic measurements of cholesterol in macrophages exposed to HDLs. Compared with HDL(Control), HDL(CKD) and HDL(ESRD) heightened the cytokine response and disrupted macrophage chemotaxis. HDL(Control) reduced endothelial expression of ICAM-1, VCAM-1, E-selectin, whereas HDL(CKD) and HDL(ESRD) were less effective and showed reduced capacity to protect endothelial cells against monocyte adhesion. Compared with a dramatically enhanced endothelial proliferation following injurious stimulus by HDL(Control), neither HDL(CKD) nor HDL(ESRD) caused proliferative effects. HDLs of all three groups were equally protective against apoptosis assessed by flow cytometry and cleaved caspase-3 activity. Compared to HDL(Control), HDL(CKD) and HDL(ESRD) trended toward reduced capacity as cholesterol acceptors. CKD in children impairs HDL function. Even in the absence of long-standing and concomitant risk factors, CKD alters specific HDL functions linked to control of inflammation and endothelial responses.