EMAIL THIS PAGE TO A FRIEND
Environmental Pollution (Barking, Essex : 1987)
Polybrominated diphenyl ethers in human placenta associated with neonatal physiological development at a typical e-waste recycling area in China.
PMID 25468211