EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Identification of odorants in frankincense (Boswellia sacra Flueck.) by aroma extract dilution analysis and two-dimensional gas chromatography-mass spectrometry/olfactometry.
PMID 25468535
Abstract
Frankincense has been known, traded and used throughout the ages for its exceptional aroma properties, and is still commonly used in both secular and religious settings to convey a pleasant odor. Surprisingly, the odoriferous principle(s) underlying its unique odor profile have never been published. In this study, resin samples of Boswellia sacra Flueck. from both Somalia and Oman were investigated by aroma extract dilution analysis. In a comprehensive, odor-activity guided approach both chemo-analytical and human-sensory parameters were used to identify odor active constituents of the volatile fraction of B. sacra. Among the key odorants found were α-pinene, β-myrcene, linalool, p-cresol and two unidentified sesquiterpenoids. Overall, a total of 23 odorants were detected and analyzed by gas chromatography-olfactometry and heart-cut two-dimensional gas chromatography-mass spectrometry/olfactometry. The majority of the identified odorant compounds were oxygenated monoterpenes, along with some relevant mono- and sesquiterpenes and only one diterpenoid substance. Several of these compounds were reported here for the first time as odorous constituents in B. sacra. Identifying bioactive compounds might support a better understanding with regard to the potential benefits of frankincense, for example in aromatherapy or ecclesial settings.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62139
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
C10H18O
74856
(−)-Linalool, analytical standard
C10H18O
183164
(R)-(+)-Limonene, 97%
C10H16
62118
(R)-(+)-Limonene, analytical standard
C10H16
W263303
(R)-(+)-Limonene, ≥93%
C10H16
62122
(R)-(+)-Limonene, technical, ~90% (sum of enantiomers, GC)
C10H16
00590590
(R)-(+)-Limonene, primary reference standard
C10H16
62120
(R)-(+)-Limonene, purum, ≥96.0% (sum of enantiomers, GC)
C10H16
00020590
1,8-Cineole, primary reference standard
C10H18O
M29355
2-Methylanisole, 99%
C8H10O
W268003
2-Methylanisole, ≥99%, FG
C8H10O
442391
3-Methylphenol, analytical standard
C7H8O
148091
4-Methylanisole, 99%
C8H10O
W268100
4-Methylanisole, ≥99%, FCC, FG
C8H10O
Y0000176
Cineole, European Pharmacopoeia (EP) Reference Standard
C10H18O
306886
Ethyl 2-methylbutyrate, 99%
C7H14O2
W244317
Ethyl 2-methylbutyrate, natural, mixture of enantiomers, ≥98%, FCC, FG
C7H14O2
W244309
Ethyl 2-methylbutyrate, ≥98%, FCC, FG
C7H14O2
112283
Ethyl isovalerate, 98%
C7H14O2
W246301
Ethyl isovalerate, ≥98%, FCC, FG
C7H14O2
W246328
Ethyl isovalerate, natural, ≥98%, FCC, FG
C7H14O2
C80601
Eucalyptol, 99%
C10H18O
W246506
Eucalyptol, natural, ≥99%, FCC, FG
C10H18O
29210
Eucalyptol, analytical standard
C10H18O
95656
Eucalyptol, certified reference material, TraceCERT®
C10H18O
L2602
Linalool, 97%
C10H18O
51782
Linalool, analytical standard
C10H18O
W263508
Linalool, ≥97%, FCC, FG
C10H18O
C85727
m-Cresol, 99%
C7H8O
65996
m-Cresol, analytical standard
C7H8O