EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Indole alkaloids from the Marquesan plant Rauvolfia nukuhivensis and their effects on ion channels.
PMID 25468537
Abstract
In addition to the already reported nukuhivensiums 1 and 2, 11 indole alkaloids were isolated from the bark of the plant Rauvolfia nukuhivensis, growing in the Marquesas archipelago. The known sandwicine (3), isosandwicine (4), spegatrine (8), lochneram (9), flavopereirine (13) have been found in this plant together with the norsandwicine (5), isonorsandwicine (6), Nb-methylisosandwicine (7), 10-methoxypanarine (10), nortueiaoine (11), tueiaoine (12). The structure elucidation was performed on the basis of a deep exploration of the NMR and HRESIMS data as well as comparison with literature data for similar compounds. Norsandwicine, 10-methoxypanarine, tueiaoine, and more importantly nukuhivensiums, were shown to significantly induce a reduction of IKr amplitude (HERG current). Molecular modelling through docking was performed in order to illustrate this result.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93595
Trypan Blue solution, 0.4%, for microscopy
C34H24N6Na4O14S4