EMAIL THIS PAGE TO A FRIEND
BMJ open diabetes research & care
Two novel type 2 diabetes loci revealed through integration of TCF7L2 DNA occupancy and SNP association data.
PMID 25469308
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-511
Anti-TCF-4 Antibody, clone 6H5-3, clone 6H5-3, Upstate®, from mouse