EMAIL THIS PAGE TO A FRIEND
Acta paediatrica (Oslo, Norway : 1992)
The endocrine disruptor bisphenol A may play a role in the aetiopathogenesis of polycystic ovary syndrome in adolescent girls.
PMID 25469562
Abstract
Experimental in vitro studies have shown that bisphenol A affects steroidogenesis, folliculogenesis and ovarian morphology. The aim of this study was to investigate the role of the endocrine disruptor bisphenol A in the aetiopathogenesis of polycystic ovary syndrome (PCOS) in adolescents and its relationship with metabolic parameters, insulin resistance and obesity in this population. A total of 112 girls with PCOS and 61 controls between 13 and 19 years of age were enrolled in the study. Serum bisphenol A levels were measured by high-pressure liquid chromatography. An oral glucose tolerance test was also performed. Adolescents with PCOS had markedly increased serum bisphenol A levels (mean: 1.1 ng/mL 95% CI: 1.0-1.2) than controls (mean: 0.8 ng/mL 95% CI: 0.6-0.9, p = 0.001). When we compared the subgroups according to obesity, the main factor determining the significant increase in bisphenol A was the presence of PCOS, but not obesity (p = 0.029). Bisphenol A was significantly correlated with total testosterone (r = 0.52), free testosterone (r = 0.44), dehydroepiandrosterone sulphate (r = 0.37) and Ferriman-Gallwey score (r = 0.43) (p < 0.05). Adolescents with PCOS had higher serum bisphenol A levels than controls, independent of obesity. Bisphenol A concentrations were significantly correlated with androgen levels, leading us to consider that bisphenol A might play a role in the aetiopathogenesis of PCOS in adolescents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42088
Bisphenol A, certified reference material, TraceCERT®
C15H16O2