PloS one
Fast and effective photodynamic inactivation of multiresistant bacteria by cationic riboflavin derivatives.
PMID 25469700
Abstract
Photodynamic inactivation of bacteria (PIB) proves to be an additional method to kill pathogenic bacteria. PIB requires photosensitizer molecules that effectively generate reactive oxygen species like singlet oxygen when exposed to visible light. To allow a broad application in medicine, photosensitizers should be safe when applied in humans. Substances like vitamin B2, which are most likely safe, are known to produce singlet oxygen upon irradiation. In the present study, we added positive charges to flavin derivatives to enable attachment of these molecules to the negatively charged surface of bacteria. Two of the synthesized flavin derivatives showed a high quantum yield of singlet oxygen of approximately 75%. Multidrug resistant bacteria like MRSA (Methicillin resistant Staphylococcus aureus), EHEC (enterohemorrhagic Escherichia coli), Pseudomonas aeruginosa, and Acinetobacter baumannii were incubated with these flavin derivatives in vitro and were subsequently irradiated with visible light for seconds only. Singlet oxygen production in bacteria was proved by detecting its luminescence at 1270 nm. After irradiation, the number of viable bacteria decreased up to 6 log10 steps depending on the concentration of the flavin derivatives and the light dosimetry. The bactericidal effect of PIB was independent of the bacterial type and the corresponding antibiotic resistance pattern. In contrast, the photosensitizer concentration and light parameters used for bacteria killing did not affect cell viability of human keratinocytes (therapeutic window). Multiresistant bacteria can be safely and effectively killed by a combination of modified vitamin B2 molecules, oxygen and visible light, whereas normal skin cells survive. Further work will include these new photosensitizers for topical application to decolonize bacteria from skin and mucosa.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R4500
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
C17H20N4O6
R7649
(−)-Riboflavin, meets USP testing specifications
C17H20N4O6
R9504
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
C17H20N4O6
151874
Dimethyl sulfoxide-d6, 99.9 atom % D
C2D6OS
296147
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
156914
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
C2D6OS
417939
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
185965
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
C2D6OS
570672
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
C2D6OS
716731
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
C2D6OS
453323
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
C2D6OS
424005
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
C2D6OS
417920
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 1 % (v/v) TMS
C2D6OS
522120
Dimethyl sulfoxide-d6, 99.9 atom % D
C2D6OS
236926
Dimethyl sulfoxide-d6, 99.9 atom % D
C2D6OS
423653
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
574341
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
C2D6OS
PHR1054
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
C17H20N4O6
1603006
Riboflavin, United States Pharmacopeia (USP) Reference Standard
C17H20N4O6
R0600000
Riboflavin, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6
47861
Riboflavin (B2), analytical standard
C17H20N4O6
Y0000757
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6