EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Discovery of MK-8970: an acetal carbonate prodrug of raltegravir with enhanced colonic absorption.
PMID 25469982
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87920
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
C4H12Si