PloS one
Naturally occurring alpha-synuclein autoantibodies in Parkinson's disease: sources of (error) variance in biomarker assays.
PMID 25470145
Abstract
Alpha-synuclein (α-Syn) plays a pivotal role in the pathophysiology of Parkinson's disease (PD), which can partly be modulated by innate and adaptive immune functions, and vice versa. Here, naturally occurring α-Syn autoantibodies (α-Syn-nAbs) may be effective against α-Syn pathoetiology and may serve as a PD biomarker. However, serum and cerebrospinal fluid α-Syn-nAbs levels still lack consistent evidence as required for a reliable PD biomarker. Serum and cerebrospinal fluid α-Syn-nAbs levels of 66 PD patients and 69 healthy controls were assessed using a validated ELISA assay. Moreover, potential sources of error variance including unspecific ELISA background signals, free serum hemoglobin concentrations, α-Syn plate coating procedures, and differences in α-Syn-nAbs standards, were investigated. PD patients and controls did not differ in serum (p = .49) nor cerebrospinal fluid (p = .29) α-Syn-nAbs levels. Interestingly, free serum hemoglobin concentrations were negatively correlated with α-Syn-nAbs levels in controls (Spearman ρ = -.41, p<.001), but not in PD patients (ρ = .16, p = .21). ELISA α-Syn plate coating procedures impacted inter-assay variability (same day coating: 8-16%; coating on different days: 16-58%). α-Syn-nAbs standards from different purification batches differed regarding optical density measured in ELISAs suggesting differences in α-Syn affinity. While α-Syn-nAbs levels may represent a potential PD biomarker, several methodological issues have to be considered to increase reproducibility of α-Syn-nAbs findings. Further studies using standardized protocols minimizing sources of error variance may be necessary to establish a reliable PD α-Syn-nAbs biomarker.