EMAIL THIS PAGE TO A FRIEND
Journal of Neuropathology and Experimental Neurology
Neuroinflammation and virus replication in the spinal cord of simian immunodeficiency virus-infected macaques.
PMID 25470348