EMAIL THIS PAGE TO A FRIEND
Pharmacology
Chemerin induces insulin resistance in rat cardiomyocytes in part through the ERK1/2 signaling pathway.
PMID 25471554
Abstract
Chemerin is a novel adipokine that is closely associated with cardiovascular diseases and glucose homeostasis. This study aimed to investigate the effects of chemerin on insulin resistance in rat cardiomyocytes. Rat cardiomyocytes were treated with high concentrations of glucose and tumor necrosis factor-alpha (TNF-α), and chemerin and chemokine-like receptor 1 (CMKLR1) were measured by Western blot analysis. Then, the cardiomyocytes were treated with chemerin and insulin. Glucose uptake was evaluated using a fluorescence microplate reader. Western blot analysis was used to evaluate the phosphorylation of Akt, insulin receptor substrate-1, p38 mitogen-activated protein kinase (MAPK), as well as extracellular signal-regulated kinase (ERK)1/2. Chemerin and CMKLR1 were found to be expressed in rat cardiomyocytes. Pretreatment with chemerin caused decreases in glucose uptake and phosphorylation of Akt in insulin-stimulated cardiomyocytes. Furthermore, chemerin activated the phosphorylation of p38 MAPK and ERK1/2 in insulin-stimulated cardiomyocytes. Inhibition of ERK partially rescued chemerin-induced insulin resistance. Chemerin is a novel adipokine that induces insulin resistance in rat cardiomyocytes in part through the ERK1/2 pathway.