EMAIL THIS PAGE TO A FRIEND
Diabetologia
Protection from diabetes-induced atherosclerosis and renal disease by D-carnosine-octylester: effects of early vs late inhibition of advanced glycation end-products in Apoe-null mice.
PMID 25471794