EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Sodium ion binding pocket mutations and adenosine A2A receptor function.
PMID 25473121
Abstract
Recently we identified a sodium ion binding pocket in a high-resolution structure of the human adenosine A2A receptor. In the present study we explored this binding site through site-directed mutagenesis and molecular dynamics simulations. Amino acids in the pocket were mutated to alanine, and their influence on agonist and antagonist affinity, allosterism by sodium ions and amilorides, and receptor functionality was explored. Mutation of the polar residues in the Na(+) pocket were shown to either abrogate (D52A(2.50) and N284A(7.49)) or reduce (S91A(3.39), W246A(6.48), and N280A(7.45)) the negative allosteric effect of sodium ions on agonist binding. Mutations D52A(2.50) and N284A(7.49) completely abolished receptor signaling, whereas mutations S91A(3.39) and N280A(7.45) elevated basal activity and mutations S91A(3.39), W246A(6.48), and N280A(7.45) decreased agonist-stimulated receptor signaling. In molecular dynamics simulations D52A(2.50) directly affected the mobility of sodium ions, which readily migrated to another pocket formed by Glu13(1.39) and His278(7.43). The D52A(2.50) mutation also decreased the potency of amiloride with respect to ligand displacement but did not change orthosteric ligand affinity. In contrast, W246A(6.48) increased some of the allosteric effects of sodium ions and amiloride, whereas orthosteric ligand binding was decreased. These new findings suggest that the sodium ion in the allosteric binding pocket not only impacts ligand affinity but also plays a vital role in receptor signaling. Because the sodium ion binding pocket is highly conserved in other class A G protein-coupled receptors, our findings may have a general relevance for these receptors and may guide the design of novel synthetic allosteric modulators or bitopic ligands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2387
5′-(N-Ethylcarboxamido)adenosine, powder
C12H16N6O4
A9561
5-(N,N-Hexamethylene)amiloride
C12H18ClN7O