EMAIL THIS PAGE TO A FRIEND
Journal of Water and Health
Water used to moisten vegetables is a source of Escherichia coli and protozoan parasite contamination at markets in Hanoi, Vietnam.
PMID 25473999