EMAIL THIS PAGE TO A FRIEND
Oncotarget
Expansion of CTCs from early stage lung cancer patients using a microfluidic co-culture model.
PMID 25474037
Abstract
The potential utility of circulating tumor cells (CTCs) to guide clinical care in oncology patients has gained momentum with emerging micro- and nanotechnologies. Establishing the role of CTCs in tumor progression and metastasis depends both on enumeration and on obtaining sufficient numbers of CTCs for downstream assays. The numbers of CTCs are few in early stages of cancer, limiting detailed molecular characterization. Recent attempts in the literature to culture CTCs isolated from metastatic patients using monoculture have had limited success rates of less than 20%. Herein, we have developed a novel in-situ capture and culture methodology for ex-vivo expansion of CTCs using a three dimensional co-culture model, simulating a tumor microenvironment to support tumor development. We have successfully expanded CTCs isolated from 14 of 19 early stage lung cancer patients. Expanded lung CTCs carried mutations of the TP53 gene identical to those observed in the matched primary tumors. Next-generation sequencing further revealed additional matched mutations between primary tumor and CTCs of cancer-related genes. This strategy sets the stage to further characterize the biology of CTCs derived from patients with early lung cancers, thereby leading to a better understanding of these putative drivers of metastasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63175
4-Maleimidobutyric acid N-hydroxysuccinimide ester, ≥98.0% (HPLC)
C12H12N2O6