EMAIL THIS PAGE TO A FRIEND
PLoS Neglected Tropical Diseases
Leishmania donovani infection enhances lateral mobility of macrophage membrane protein which is reversed by liposomal cholesterol.
PMID 25474261