EMAIL THIS PAGE TO A FRIEND
PLoS Neglected Tropical Diseases
Immunizing adult female mice with a TcpA-A2-CTB chimera provides a high level of protection for their pups in the infant mouse model of cholera.
PMID 25474636