EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Protic ionic liquids (PILs) nanostructure and physicochemical properties: development of high-throughput methodology for PIL creation and property screens.
PMID 25475718
Abstract
A high-throughput approach was developed in order to prepare and dry a series of protic ionic liquids (PILs) from 48 Brønsted acid-base combinations. Many combinations comprised an alkyl carboxylic acid paired with an alkyl amine. Visual screens were developed to identify which acid-base combinations formed PILs, and of those, which PILs were likely to have high surface tensions, low viscosities, and low melting points. The surface tension screen was validated through pendant drop surface tension measurements. Karl Fischer coulometric titration was used to obtain the water contents, and it was noted that there is a considerable difference in the drying rate throughout this series of PILs. It was observed that an octyl chain present on either the cation or anion was detrimental to the formation of a PIL with a low melting point, and instead increased the likelihood of a gel or solid forming. The nanostructure of the PILs was determined, using synchrotron small and wide angle X-ray scattering (SAXS/WAXS), to consist of polar and non-polar domains, with the alkyl chains on the cation and anion intercalating. The results indicate that both the alkyl chain on the cation and/or anion contribute to the correlation distance, for the intermediate range order, with the expectation that there is charge alternation of the ions in the polar region. The maximum correlation distance was observed when there was an alkyl chain present on only one ion. This correlation distance could be significantly reduced by varying the alkyl chain length present on the other ion, which was attributed to increased disorder and interdigitation of chains, and to toe-to-toe alignment of the chains. To the best of our knowledge this is the first PIL report into the effect of having an alkyl chain present on both the cation and the anion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
171409
Amylamine, 99%
C5H13N
68518
Amylamine, analytical standard
C5H13N
W424201
Amylamine, ≥99%, FG
C5H13N
473316
Amylamines, mixture of isomers, ≥98%
C5H13N
C0426000
Caprylic acid, European Pharmacopoeia (EP) Reference Standard
C8H16O2
PHR1202
Caprylic Acid (Octanoic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
C8H16O2
02946
Ethylamine, anhydrous, ≥99.5%
C2H7N
301264
Ethylamine, 97%
C2H7N
724033
Ethylamine, 99.0%
C2H7N
395064
Ethylamine solution, 2.0 M in methanol
C2H7N
395072
Ethylamine solution, 2.0 M in THF
C2H7N
471208
Ethylamine solution, 66.0-72.0% in H2O
C2H7N
75190
Heptanoic acid, ≥99.0% (GC)
C7H14O2
W334812
Heptanoic acid, natural, FG
C7H14O2
43858
Heptanoic acid, analytical standard
C7H14O2
146870
Heptanoic acid, 96%
C7H14O2
W334804
Heptanoic acid, 97%, FG
C7H14O2
153745
Hexanoic acid, ≥99%
C6H12O2
W255904
Hexanoic acid, ≥98%, FCC, FG
C6H12O2
W255912
Hexanoic acid, natural, ≥98%, FCC
C6H12O2
21529
Hexanoic acid, analytical standard
C6H12O2
21530
Hexanoic acid, purum, ≥98.0% (GC)
C6H12O2
50803
Hexylamine, analytical standard
C6H15N
219703
Hexylamine, 99%
C6H15N
W279900
Octanoic acid, ≥98%, FG
C8H16O2
W279927
Octanoic acid, natural, ≥98%, FG
C8H16O2
O3907
Octanoic acid, ≥98%
C8H16O2
C2875
Octanoic acid, ≥99%
C8H16O2
21639
Octanoic acid, analytical standard
C8H16O2
74989
Octylamine, analytical standard
C8H19N