EMAIL THIS PAGE TO A FRIEND
Biochemical and Biophysical Research Communications
Methylation-associated silencing of miR-495 inhibit the migration and invasion of human gastric cancer cells by directly targeting PRL-3.
PMID 25475733