EMAIL THIS PAGE TO A FRIEND
Journal of animal physiology and animal nutrition
Effect of an extruded pea or rice diet on postprandial insulin and cardiovascular responses in dogs.
PMID 25475789
Abstract
Peas are increasing in popularity as a source of carbohydrate, protein and fibre in extruded canine diets. The aim of this study was to test the health effects of two canine diets with identical macronutrient profiles, but containing either yellow field peas or white rice as the carbohydrate source on metabolism, cardiovascular outcomes and adiposity. First, the acute glycemic, insulinemic and cardiovascular responses to the pea- or rice-based diets were determined in normal weight beagles (nxa0=xa07 dogs). The glycemic index did not differ between the pea diet (56xa0±xa012) and rice diet (63xa0±xa09). Next, obese beagles (nxa0=xa09) were fed the yellow field pea diet or white rice diet ad libitum for 12xa0weeks in a crossover study. Adiposity (measured using computed tomography), metabolic (oral glucose tolerance test, plasma leptin, adiponectin, C-reactive protein) and cardiovascular assessments (echocardiography and blood pressure) were performed before and after each crossover study period. After 12xa0weeks on each diet, peak insulin (pxa0=xa00.05) and area under the curve (AUC) for insulin after a 10xa0g oral glucose tolerance test (pxa0=xa00.05) were lower with the pea than the rice diet. Diet did not show a significant effect on body weight, fat distribution, cardiovascular variables, adiponectin or leptin. In conclusion, a diet containing yellow field peas reduced the postprandial insulin response after glucose challenge in dogs despite continued obesity, indicating improved metabolic health.