EMAIL THIS PAGE TO A FRIEND
Malaria journal
Quality and stability of artemether-lumefantrine stored under ambient conditions in rural Mali.
PMID 25476144
Abstract
The quality and stability of anti-malarial drugs in the Global South has long been of significant concern. Drug quality can be affected by poor or fraudulent manufacturing processes, while drug stability is affected by temperature and humidity. Knowledge of drug quality and stability is often the unique contribution of pharmacists volunteering on short-term medical mission trips. To determine the quality and stability of artemether-lumefantrine 20/120 mg under ambient storage conditions in rural Mali. One unopened blister pack of artemether-lumefantrine 20/120 mg (IPCA Laboratories, Mumbai) was stored under ambient conditions in a warehouse in a Malian village for one year. A second pack from the same lot number was stored under temperature and humidity controlled conditions in a university laboratory. The active ingredients of tablets from both packages were analysed using thin layer chromatography, nuclear magnetic resonance and infrared spectroscopy. The IPCA samples were referenced for drug identity and content to an identical American made product (Coartem(®), Novartis Pharmaceuticals). Thin layer chromatographs, nuclear magnetic resonance and infrared spectroscopy results were identical for both IPCA samples and the reference product. The IPCA products contained the same drugs in the same amount as on their package label and were identical to the reference product. It is concluded that they were of good quality. Spectroscopy results demonstrate the sample stored in Mali was stable for one year. Pharmacists volunteering on medical mission trips may dispense this product with confidence. At the end of a mission trip, pharmacists may store left over artemether-lumefantrine under ambient conditions for up to one year without concern for significant degradation of the active ingredients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5420
Lumefantrine
C30H32Cl3NO