EMAIL THIS PAGE TO A FRIEND
Talanta
A novel approach for high sensitive determination of sulfur mustard by derivatization and isotope-dilution LC-MS/MS analysis.
PMID 25476305
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241776
Potassium thioacetate, 98%
C2H3KOS