EMAIL THIS PAGE TO A FRIEND
Talanta
Vortex-assisted low density solvent liquid-liquid microextraction and salt-induced demulsification coupled to high performance liquid chromatography for the determination of five organophosphorus pesticide residues in fruits.
PMID 25476376
Abstract
A simple and rapid microextraction method, vortex-assisted low density solvent liquid-liquid microextraction and salt-induced demulsification (VLLME-SID) coupled to high performance liquid chromatography (HPLC) was developed for the determination of organophosphorus pesticide (OPP) residues in fruits. The studied OPPs were azinphos-methyl, parathion-methyl, fenitrothion, diazinon and chlorpyrifos. For VLLME-SID, a mixture of low density solvents (1-dodecanol and hexane) was used as the extraction solvent under vortex agitation for enhancing dispersion. After complete dispersion, the emulsion was formed and the OPPs were extracted into extraction solvent droplets. Then, the emulsion was quickly broken up into two clear phases after the addition of AlCl3 as a demulsifier. Centrifugation was not required in this procedure. Under the optimal conditions, high enrichment factors (180-282), low limit of detections (LODs) (0.05-1 ng mL(-1)) and good precision (RSD≤7% for retention time and peak area) were obtained. The proposed method was successfully applied to the analysis of OPP residues in fruit samples (watermelon, grape, and cantaloupe). The LODs for samples were in the range 0.0006-0.0015 mg kg(-1) which are below the established EU-MRLs (0.01-0.3 mg kg(-1)). Good recoveries were also obtained (80-104%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44095
1-Dodecanol, Selectophore, ≥98.0%
C12H26O
443816
1-Dodecanol, ACS reagent, ≥98.0%
C12H26O
126799
1-Dodecanol, reagent grade, 98%
C12H26O
75544
1-Dodecanol, analytical standard
C12H26O
U1001
1-Undecanol, 99%
C11H24O
D221503
2-Dodecanol, 99%
C12H26O
A5508
Adenosine 5′-phosphosulfate sodium salt, ≥85%
C10H14N5O10PS
03382
Aluminum atomic spectroscopy standard concentrate 10.00 g Al, 10.00 g/L, for 1 l standard solution, analytical standard
AlCl3
563919
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
AlCl3
294713
Aluminum chloride, 99.99% trace metals basis
AlCl3
449598
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
AlCl3
249882
Aluminum chloride solution, 1.0 M in nitrobenzene
AlCl3
06154
Aluminum standard for AAS, analytical standard, traceable to BAM
AlCl3
45333
Azinphos-methyl, PESTANAL®, analytical standard
C10H12N3O3PS2
45395
Chlorpyrifos, PESTANAL®, analytical standard
C9H11Cl3NO3PS
94114
Chlorpyrifos, certified reference material, TraceCERT®
C9H11Cl3NO3PS
45428
Diazinon, PESTANAL®, analytical standard
C12H21N2O3PS
68486
Diazinon, certified reference material, TraceCERT®
C12H21N2O3PS
45487
Fenitrothion, PESTANAL®, analytical standard
C9H12NO5PS
W261718
Lauryl alcohol, ≥98%, FG
C12H26O
W309702
Undecyl alcohol, 97%, FG
C11H24O