EMAIL THIS PAGE TO A FRIEND
Malaria Journal
The effect of UGTs polymorphism on the auto-induction phase II metabolism-mediated pharmacokinetics of dihydroartemisinin in healthy Chinese subjects after oral administration of a fixed combination of dihydroartemisinin-piperaquine.
PMID 25476790