EMAIL THIS PAGE TO A FRIEND
Investigational New Drugs
PBI-05204, a supercritical CO? extract of Nerium oleander, inhibits growth of human pancreatic cancer via targeting the PI3K/mTOR pathway.
PMID 25476893
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06069
Oleandrin, analytical standard
C32H48O9