EMAIL THIS PAGE TO A FRIEND
Food & Function
Utilisation of the isobole methodology to study dietary peptide-drug and peptide-peptide interactive effects on dipeptidyl peptidase IV (DPP-IV) inhibition.
PMID 25477187
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9759
Ile-Pro-Ile, ≥97% (HPLC)
C17H31N3O4