EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Gastrointestinal and Liver Physiology
In vitro evidence that phosphatidylcholine protects against indomethacin/bile acid-induced injury to cells.
PMID 25477376