EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Build-up and wash-off dynamics of atmospherically derived Cu, Pb, Zn and TSS in stormwater runoff as a function of meteorological characteristics.
PMID 25478658
Abstract
Atmospheric pollutants deposited on impermeable surfaces can be an important source of pollutants to stormwater runoff; however, modelling atmospheric pollutant loads in runoff has rarely been done, because of the challenges and uncertainties in monitoring their contribution. To overcome this, impermeable concrete boards (≈ 1m(2)) were deployed for 11 months in different locations within an urban area (industrial, residential and airside) throughout Christchurch, New Zealand, to capture spatially distributed atmospheric deposition loads in runoff over varying meteorological conditions. Runoff was analysed for total and dissolved Cu, Zn, Pb, and total suspended solids (TSS). Mixed-effect regression models were developed to simulate atmospheric pollutant loads in stormwater runoff. In addition, the models were used to explain the influence of different meteorological characteristics (e.g. antecedent dry days and rain depth) on pollutant build-up and wash-off dynamics. The models predicted approximately 53% to 69% of the variation in pollutant loads and were successful in predicting pollutant-load trends over time which can be useful for general stormwater planning processes. Results from the models illustrated the importance of antecedent dry days on pollutant build-up. Furthermore, results indicated that peak rainfall intensity and rain duration had a significant relationship with TSS and total Pb, whereas, rain depth had a significant relationship with total Cu and total Zn. This suggested that the pollutant speciation phase plays an important role in surface wash-off. Rain intensity and duration had a greater influence when the pollutants were predominantly in their particulate phase. Conversely, rain depth exerted a greater influence when a high fraction of the pollutants were predominantly in their dissolved phase. For all pollutants, the models were represented by a log-arctan relationship for pollutant build-up and a log-log relationship for pollutant wash-off. The modelling approach enables the site-specific relationships between individual pollutants and rainfall characteristics to be investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR058
Continuous cast copper (O), BCR® certified Reference Material, rod
Cu
311405
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Cu
203122
Copper, powder, 99.999% trace metals basis
Cu
266086
Copper, powder, <425 μm, 99.5% trace metals basis
Cu
326453
Copper, powder (spheroidal), 10-25 μm, 98%
Cu
357456
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Cu
207780
Copper, powder, <75 μm, 99%
Cu
349216
Copper, wire, diam. 1.0 mm, ≥99.9%
Cu
61139
Copper, turnings, purum p.a., ≥99.0%
Cu
349208
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Cu
349178
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Cu
326488
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Cu
12816
Copper, foil, ≥99.8% (complexometric)
Cu
254177
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Cu
774111
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Cu
774103
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Cu
31284
Copper, electrolytic, Analytical Reagent, Reag. Ph. Eur., ≥99.9% (complexometric), wire
Cu
520381
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Cu
349151
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Cu
223409
Copper, shot, −3-+14 mesh, 99%
Cu
266744
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Cu
326410
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Cu
349224
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Cu
326445
Copper, bars (random sizes), ≥99.999% trace metals basis
Cu
326429
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Cu
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF86675439
Copper, wire reel, 10m, diameter 0.01mm, hard, 99.99%
Cu
GF99677165
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Cu
GF66556491
Copper, mesh, 100x100mm, nominal aperture 0.14mm, thickness 0.25mm, wire diameter 0.115mm, 100x100 wires/inch, open area 30.3%, plain weave mesh
Cu
GF93417105
Copper, wire reel, 100m, diameter 1.0mm, as drawn, 99.98+%
Cu