EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
Evaluation and comparison of a new DOTA and DTPA-bombesin agonist in vitro and in vivo in low and high GRPR expressing prostate and breast tumor models.
PMID 25479439
Abstract
We evaluated and compared a new bombesin analog [Tyr-Gly5, Nle(14)]-BBN(6-14) conjugated to DOTA or DTPA and radiolabeled with In-111 in low and high GRPR expressing tumor models. Both peptides were radiolabeled with high radiochemical purity and specific activity. In vitro assays on T-47D, LNCaP and PC-3 cells showed that the affinity of peptides is similar and a higher binding and internalization of DOTA-peptide to PC-3 cells was observed. Both peptides could target PC-3 and LNCaP tumors in vivo and both tumor types could be visualized by microSPECT/CT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32319
Diethylenetriaminepentaacetic acid, for complexometry, ≥99.0%
C14H23N3O10