EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Two minor chalcone acetylglycosides from the roots extract of Glycyrrhiza uralensis.
PMID 25479707
Abstract
An extensive phytochemical investigation on the roots of Glycyrrhiza uralensis led to the isolation of two new minor chalcone acetylglycosides, i.e., 6″-O-acetylisoliquiritin (1) and 6″-O-acetylneoisoliquiritin (2), including 16 kinds of known constituents (3-18) of flavonoids, chalcones and triterpene saponins. The chemical structures of 1 and 2 were established by spectroscopic analyses of them, particularly by the aid of two-dimensional NMR experiments, COSY, DEPT, HSQC and HMBC. Some isolated components except 1 and 2 exhibited significant inhibitory effects on the proliferation of cultured tumor cell lines, such as A549, SK-OV-3, A-498, and HCD15, in vitro.