EMAIL THIS PAGE TO A FRIEND
Archives of Environmental Contamination and Toxicology
Concentrations, gas-particle partitioning, and seasonal variations of polycyclic aromatic hydrocarbons at four sites in Turkey.
PMID 25480127
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
451819
Acenaphthene-d10, 99 atom % D
C12D10
442432
Acenaphthene-d10, analytical standard
C12D10
416703
Acenaphthylene, 99%
C12H8
A805
Acenaphthylene, 75%
C12H8
92549
Acenaphthylene, certified reference material, TraceCERT®
C12H8
48566
Acenaphthylene, analytical standard
C12H8
B2209
Benz[a]anthracene, 99%
C18H12
48563
Benz[a]anthracene, analytical standard
C18H12
BCR271
Benz[a]anthracene, BCR® certified Reference Material
C18H12
75451
Benz[a]anthracene, certified reference material, TraceCERT®
C18H12
36956
Benz[a]anthracene solution, 100 ng/μL in cyclohexane, analytical standard
C18H12
48651
Benz[a]anthracene solution, certified reference material, 200 μg/mL in methylene chloride
C18H12
40070-U
Benz[a]anthracene solution, certified reference material, 1000 μg/mL in methanol
C18H12
35754
Chrysene, analytical standard
C18H12
245186
Chrysene, 98%
C18H12
48565-U
Chrysene, analytical standard
C18H12
BCR269
Chrysene, BCR® certified Reference Material
C18H12
94035
Chrysene, certified reference material, TraceCERT®
C18H12
128333
Fluorene, 98%
C13H10
56849
Fluorene, certified reference material, TraceCERT®
C13H10
48568
Fluorene, analytical standard
C13H10
176044
Naphthalene-d8, 99 atom % D, ≥98% (CP)
C10D8
442716
Naphthalene-d8, analytical standard
C10D8