EMAIL THIS PAGE TO A FRIEND
The International Journal of Biochemistry & Cell Biology
Involvement of BK channel in differentiation of vascular smooth muscle cells induced by mechanical stretch.
PMID 25481230