EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and thrombolysis
A plasmin-activatable thrombin inhibitor reduces experimental thrombosis and assists experimental thrombolysis in murine models.
PMID 25481811