EMAIL THIS PAGE TO A FRIEND
Cell cycle (Georgetown, Tex.)
Involvement of de novo synthesized palmitate and mitochondrial EGFR in EGF induced mitochondrial fusion of cancer cells.
PMID 25483192
Abstract
Increased expressions of fatty acid synthase (FASN) and epidermal growth factor receptor (EGFR) are common in cancer cells. De novo synthesis of palmitate by FASN is critical for the survival of cancer cells via mechanisms independent of its role as an energy substrate. Besides the plasma membrane and the nucleus, EGFR can also localize at the mitochondria; however, signals that can activate mitochondrial EGFR (mtEGFR) and the functions of mtEGFR of cancer cells remain unknown. The present study characterizes mtEGFR in the mitochondria of cancer cells (prostate and breast) and reveals that mtEGFR can promote mitochondrial fusion through increasing the protein levels of fusion proteins PHB2 and OPA1. Activation of plasma membranous EGFR (pmEGFR) stimulates the de novo synthesis of palmitate through activation of FASN and ATP-citrate lyase (ACLy). In vitro kinase assay with isolated mitochondria shows that palmitate can activate mtEGFR. Inhibition of FASN blocks the mtEGFR phosphorylation and palmitoylation induced by EGF. Mutational studies show that the cysteine 797 is important for mtEGFR activation and palmitoylation. Inhibition of FASN can block EGF induced mitochondrial fusion and increased the sensitivity of prostate cancer cells to EGFR tyrosine kinase inhibitor. In conclusion, these results suggest that mtEGFR can be activated by pmEGFR through de novo synthesized palmitate to promote mitochondrial fusion and survival of cancer cells. This mechanism may serve as a novel target to improve EGFR-based cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-1050-M
4G10 Platinum, Anti-Phosphotyrosine Antibody (mouse monoclonal cocktail IgG2b), clone 4G10®, Upstate®, from mouse
SAB4300352
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
SAB4300353
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
SAB4300510
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
SAB4300472
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAB4300602
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
SAB4300454
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
GW21146 Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405746
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018530
Anti-EGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001200
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500711
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500712
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB1405796
Anti-FASN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA006461
Anti-FASN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300700
Anti-FASN antibody produced in rabbit, affinity isolated antibody
F9554 Anti-Fatty Acid Synthase antibody produced in rabbit, 1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4504579
Anti-phospho-EGFR (pSer1026) antibody produced in rabbit, affinity isolated antibody
SAB4504382
Anti-phospho-EGFR (pSer1070) antibody produced in rabbit, affinity isolated antibody
SAB4504580
Anti-phospho-EGFR (pSer1071) antibody produced in rabbit, affinity isolated antibody
SAB4504578
Anti-phospho-EGFR (pSer695) antibody produced in rabbit, affinity isolated antibody
SAB4300151
Anti-phospho-EGFR (pThr678) antibody produced in rabbit, affinity isolated antibody
SAB4503759
Anti-phospho-EGFR (pThr678) antibody produced in rabbit, affinity isolated antibody
SAB4300152
Anti-phospho-EGFR (pThr693) antibody produced in rabbit, affinity isolated antibody
SAB4503909
Anti-phospho-EGFR (pTyr1016) antibody produced in rabbit, affinity isolated antibody
SAB4504170
Anti-phospho-EGFR (pTyr1069) antibody produced in rabbit, affinity isolated antibody
SAB4300063
Anti-phospho-EGFR (pTyr1092) antibody produced in rabbit, affinity isolated antibody
SAB4504383
Anti-phospho-EGFR (pTyr1092) antibody produced in rabbit, affinity isolated antibody
SAB4300209
Anti-phospho-EGFR (pTyr1110) antibody produced in rabbit, affinity isolated antibody