EMAIL THIS PAGE TO A FRIEND
Anticancer research
Different binding to DNA of enantiomeric platinum complexes assessed by inhibition of restriction enzyme activity.
PMID 2548438
Abstract
The ability of enantiomeric platinum complexes to block the action of selected restriction enzymes has been investigated. The complexes [PtCl2(DAC)], [PtCl2(DAB)] and [PtCl2(DAP)] (DAC = 1,2-diamminocyclohexane; DAB = 2,3-diamminobutane; DAP = 1,2-diamminopropane) exhibit a guanine-cytosine preference in accord with previous results on cis-[PtCl2(NH3)2] (cis-DDP). The extent of inhibition, however, is significantly different for the different isomers; the R,R form is more active than the others at short incubation time, as the time becomes longer, the differences among isomers level off. It also appears that cis-DDP is more active than [PtCl2(DAC)] in blocking the Cfo I enzyme, though it shows a preference for the G-X-G sequences.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
404322
Dichloro(1,2-diaminocyclohexane)platinum(II)
C6H14Cl2N2Pt