EMAIL THIS PAGE TO A FRIEND
Canadian Urological Association journal = Journal de l'Association des urologues du Canada
Effect of dutasteride in men receiving intermittent androgen ablation therapy: The AVIAS trial.
PMID 25485005
Abstract
We studied the effect of dutasteride on the length of the off-treatment period in prostate cancer patients on intermittent androgen deprivation (IAD) therapy. We conducted a randomized, placebo-controlled Phase II trial in men with localized prostate cancer and a rising prostate-specific antigen (PSA) level post-primary treatment. Patients were randomized to dutasteride (0.5 mg/day) or placebo. All patients received androgen deprivation therapy (ADT), which was stopped at month 9 if the PSA level was <1.0 ng/mL. ADT was resumed when PSA increased to ≥5.0 ng/mL. End points included time off treatment, PSA nadir after 9 months of ADT, serum testosterone and dihydrotestosterone levels, and time to castrate-resistant prostate cancer (rising PSA while testosterone levels remain <50 ng/mL). There were 87 evaluable patients: 49 dutasteride, 38 placebo. In total, 80 patients completed one treatment cycle: 45 dutasteride, 35 placebo. The median time off treatment for patients reaching ≥5 ng/mL was 18.6 and 16.7 months for dutasteride and placebo, respectively (p = 0.7600). The median PSA nadir at 9 months was 0.1 and 0.075 ng/mL, respectively (p = 0.4486). There were no cases of androgen-independent prostate cancer. Our study limitations include its short duration with only one treatment cycle evaluated. This small-scale Phase II randomized controlled trial showed no benefit to the addition of dutasteride to an IAD regimen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001598
Dutasteride, European Pharmacopoeia (EP) Reference Standard
C27H30F6N2O2