EMAIL THIS PAGE TO A FRIEND
Biological engineering transactions
Non-Instrumented Nucleic Acid Amplification (NINA) for Rapid Detection of
PMID 25485176
Abstract
We report on the use of a non-instrumented device for the implementation of a loop-mediated amplification (LAMP) based assay for the select-agent bacterial-wilt pathogen
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248568
Calcium oxide, reagent grade
CaO