EMAIL THIS PAGE TO A FRIEND
Laboratory Investigation; a Journal of Technical Methods and Pathology
Inactivation of Notch signaling reverses the Th17/Treg imbalance in cells from patients with immune thrombocytopenia.
PMID 25485537