EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Analysis of gadolinium-based contrast agents in tap water with a new hydrophilic interaction chromatography (ZIC-cHILIC) hyphenated with inductively coupled plasma mass spectrometry.
PMID 25486924
Abstract
Hydrophilic interaction chromatography (HILIC) coupled with inductively coupled plasma mass spectrometry (ICP-MS) were optimised for the direct determination of gadolinium-based contrast agents in tap water. In comparison to our previous work, a new developed zwitterionic HILIC column (ZIC-cHILIC) was used for speciation of Gd-containing contrast agents. The limit of quantification (LOQ) for the five contrast agents Gd-BOPTA, Gd-DPTA-BMA, Gd-BT-DO3A, Gd-DOTA and Gd-DTPA are in the range of 5-12 ng Gd per litre. Additionally, a new internal standard, Pr-DOTA, was investigated to correct intensity drifts, minor and major changes in the sample volumes and possible matrix effects. With the speciation method described, tap water samples from the area of Berlin were analysed and for the first time, three Gd species, Gd-BT-DO3A, Gd-DOTA and Gd-BOPTA, were found in tap water samples at concentrations of about 10-20 ng Gd per litre. These are the same Gd species which have been previously detected predominantly in surface waters of the Berlin area.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86734
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, ≥97.0% (CHN)
C16H28N4O8 · xH2O
261173
Praseodymium, ingot, under oil, 99.9% trace rare earth metals basis
Pr
263176
Praseodymium, powder, ~40 mesh, 99.5% trace rare earth metals basis
Pr
GF48004804
Praseodymium, foil, not light tested, 50x50mm, thickness 0.01mm, as rolled, 99%
Pr
GF66110329
Praseodymium, rod, 100mm, diameter 6.35mm, cast, 99%
Pr
GF06128883
Praseodymium, rod, 50mm, diameter 6.35mm, cast, 99%
Pr
GF50132191
Praseodymium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Pr
GF66524643
Praseodymium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%
Pr
GF92630246
Praseodymium, foil, not light tested, 50x50mm, thickness 0.015mm, as rolled, 99%
Pr
GF34755857
Praseodymium, foil, not light tested, 50x50mm, thickness 0.005mm, as rolled, 99%
Pr
GF82608295
Praseodymium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Pr
GF69747621
Praseodymium, foil, not light tested, 25x25mm, thickness 0.01mm, as rolled, 99%
Pr
GF75299170
Praseodymium, foil, not light tested, 25x25mm, thickness 0.015mm, as rolled, 99%
Pr
GF96026531
Praseodymium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Pr
GF07601059
Praseodymium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Pr
GF91191191
Praseodymium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%
Pr
GF98899687
Praseodymium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%
Pr