EMAIL THIS PAGE TO A FRIEND
BMJ (Clinical research ed.)
Most of UK is now out of step with rest of Europe on drink driving limits.
PMID 25487277
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02870
Ethanol, puriss. p.a., ACS reagent, prima fine spirit, without additive, F15 o1, ~96%
C2H6O
24102
Ethanol, absolute, ≥99.8% (GC)
C2H6O
02890
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
C2H6O
E-034
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-040
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
C2H6O
E-031
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-038
Ethanol-100 (5 ampules/kit), 100 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
E-041
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-056
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-043
Ethanol-20 (5 ampules/kit), 20 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
E-032
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-039
Ethanol-200 (5 ampules/kit), 200 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
E-035
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-033
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-045
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-036
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-044
Ethanol-400 (5 ampules/kit), 400 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
E-029
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-053
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-030
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-037
Ethanol-80, 80 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
187380 Ethyl alcohol, denatured, reagent grade
E7023
Ethyl alcohol, Pure, 200 proof, for molecular biology
C2H6O
459844
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
C2H6O
459828
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
C2H6O
E7148
Ethyl alcohol, Pure, 190 proof, for molecular biology
C2H6O
493546
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
C2H6O
277649
Reagent Alcohol, anhydrous, ≤0.003% water
676829
Reagent Alcohol, anhydrous, ≤0.005% water
362808
Reagent Alcohol, reagent grade