EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biology
Both high-fidelity replicative and low-fidelity Y-family polymerases are involved in DNA rereplication.
PMID 25487575
Abstract
DNA rereplication is a major form of aberrant replication that causes genomic instabilities, such as gene amplification. However, little is known about which DNA polymerases are involved in the process. Here, we report that low-fidelity Y-family polymerases (Y-Pols), Pol η, Pol ι, Pol κ, and REV1, significantly contribute to DNA synthesis during rereplication, while the replicative polymerases, Pol δ and Pol ε, play an important role in rereplication, as expected. When rereplication was induced by depletion of geminin, these polymerases were recruited to rereplication sites in human cell lines. This finding was supported by RNA interference (RNAi)-mediated knockdown of the polymerases, which suppressed rereplication induced by geminin depletion. Interestingly, epistatic analysis indicated that Y-Pols collaborate in a common pathway, independently of replicative polymerases. We also provide evidence for a catalytic role for Pol η and the involvement of Pol η and Pol κ in cyclin E-induced rereplication. Collectively, our findings indicate that, unlike normal S-phase replication, rereplication induced by geminin depletion and oncogene activation requires significant contributions of both Y-Pols and replicative polymerases. These findings offer important mechanistic insights into cancer genomic instability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32883
4-tert-Octylphenol monoethoxylate solution, 1 μg/mL in acetone, analytical standard
C16H26O2
32882
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
C16H26O2
C6891
5-Chloro-2′-deoxyuridine, thymidine analog
C9H11ClN2O5
HPA027555
Anti-POLE2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
E0396
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
C14H24N2O10
E3889
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
C14H24N2O10
E4378
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
C14H24N2O10
03777
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
C14H24N2O10
Y0000119
Hydroxycarbamide, European Pharmacopoeia (EP) Reference Standard
CH4N2O2
H8627
Hydroxyurea, 98%, powder
CH4N2O2
63031
Magnesium ion solution for ISE, 0.01 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
63042
Magnesium ion solution for ISE, 0.1 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
RDD004
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
C8H18N2O6S2
P6757
PIPES, ≥99% (titration)
C8H18N2O6S2
RES0703P-A7
PIPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O6S2
P1851
PIPES, BioPerformance Certified, suitable for cell culture, ≥99%
C8H18N2O6S2
P8203
PIPES, BioXtra, ≥99% (titration)
C8H18N2O6S2
80635
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
C8H18N2O6S2
81845
Propidium iodide, ≥94% (HPLC)
C27H34I2N4
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa
84100
Sucrose, for microbiology, ACS reagent, ≥99.0%
C12H22O11
S1600000
Sucrose, European Pharmacopoeia (EP) Reference Standard
C12H22O11
1623637
Sucrose, United States Pharmacopeia (USP) Reference Standard
C12H22O11
PHR1001
Sucrose
C12H22O11
S1174
Sucrose, analytical standard, for enzymatic assay kit SCA20
C12H22O11
47289
Sucrose, analytical standard
C12H22O11