EMAIL THIS PAGE TO A FRIEND
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine
Lycorine induces programmed necrosis in the multiple myeloma cell line ARH-77.
PMID 25487618