EMAIL THIS PAGE TO A FRIEND
Medicina (Kaunas, Lithuania)
Relationship between ACTN3 R577X polymorphism and maximal power output in elite Polish athletes.
PMID 25488167
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01070
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione, ≥98.0% (HPLC)
C10H14O3