EMAIL THIS PAGE TO A FRIEND
Nature communications
Stretchable silicon nanoribbon electronics for skin prosthesis.
PMID 25490072
Abstract
Sensory receptors in human skin transmit a wealth of tactile and thermal signals from external environments to the brain. Despite advances in our understanding of mechano- and thermosensation, replication of these unique sensory characteristics in artificial skin and prosthetics remains challenging. Recent efforts to develop smart prosthetics, which exploit rigid and/or semi-flexible pressure, strain and temperature sensors, provide promising routes for sensor-laden bionic systems, but with limited stretchability, detection range and spatio-temporal resolution. Here we demonstrate smart prosthetic skin instrumented with ultrathin, single crystalline silicon nanoribbon strain, pressure and temperature sensor arrays as well as associated humidity sensors, electroresistive heaters and stretchable multi-electrode arrays for nerve stimulation. This collection of stretchable sensors and actuators facilitate highly localized mechanical and thermal skin-like perception in response to external stimuli, thus providing unique opportunities for emerging classes of prostheses and peripheral nervous system interface technologies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
461210
Cerium, chips
Ce
261041
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Ce
263001
Cerium, powder, −40 mesh, 99.9% trace rare earth metals basis
Ce
GF97217220
Cerium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Ce
GF49222582
Cerium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Ce
GF61818363
Cerium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Ce
GF27295724
Cerium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.9%
Ce
GF97378677
Cerium, foil, light tested, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Ce
GF36157394
Cerium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Ce
GF90687695
Cerium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ce
GF39030353
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Ce
GF04444095
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Ce
GF55512244
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Ce
GF02435554
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Ce
GF35961655
Cerium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ce
GF49695047
Cerium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Ce
95660
o-Xylene, analytical standard
C8H10
PHR1312
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10
O7805
Oleylamine, technical grade, 70%
C18H37N