EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Exosome from chaperone-rich cell lysates-loaded dendritic cells produced by CELLine 1000 culture system exhibits potent immune activity.
PMID 25490390
Abstract
Dendritic cells (DCs) pulsed with exosomes can stimulate efficient cytotoxic T-lymphocyte responses and anti-tumor immunity. However, the quantity of DC-derived exosomes (DCex) obtained from various culture systems is very low, which is a significant practical issue hampering progress in this research area and needs to be addressed. Gliomas were particularly aggressive, with high morbidity and mortality, indicating that this is a form of incurable highly malignant tumor of the brain with poor prognosis. In the present study, we demonstrate that the CELLine 1000 culture system can dramatically increase the production of DCex. The morphology, phenotype and immune molecules of these DCex were found to be identical to those using traditional methods. Our researches supply a cost-effective, useful method for significantly increasing the quantity of exosomes. In addition, GL261 glioma cells were chosen to separate chaperone-rich cell lysates (CRCL). The results indicate that CRCL-GL261 cell lysates can trigger the most intense expression of immune molecules on DCex or DCs, which has important implications for the research into tumor treatment and diagnosis.