EMAIL THIS PAGE TO A FRIEND
Journal of Pharmacology and Experimental Therapeutics
Omega-3 fatty acids and other FFA4 agonists inhibit growth factor signaling in human prostate cancer cells.
PMID 25491146
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G9797
GW9508, ≥98% (HPLC)
C22H21NO3