EMAIL THIS PAGE TO A FRIEND
Indian journal of public health
Predictors of breastfeeding problems in the first postnatal week and its effect on exclusive breastfeeding rate at six months: experience in a tertiary care centre in Northern India.
PMID 25491520
Abstract
In spite of the countless benefits of breastfeeding, prevalence of exclusive breastfeeding (EBF) has been far from optimal in the developing world. Breastfeeding problems at or after 4 weeks has been reported as one among the constraints to EBF. The study aimed to determine the breastfeeding problems in the 1 st postnatal week, their predictors and impact on EBF rate at 6 months. Under a prospective cohort design, 400 mother-newborn dyads were assessed for breastfeeding problems before discharge and at 60 ± 12 h of discharge. Nearly 89% of the mother-newborn dyads had one or more BF problems before discharge. Major concern was difficulty in positioning and attaching the infant to the breast (88.5%), followed by breast and nipple problems (30.3%). BF problems continued to persist even after discharge in a significant proportion of the mothers (72.5%). The only independent predictor of BF problems in the 1 st week was the caesarean section (odds ratio: 1.9, 95% confidence interval: 1.3-3.2, P < 0.05). There was a marked improvement in the EBF status (69.5%) at 6 months, and BF problems did not predict EBF failure at 6 months.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
690651
(S)-(−)-2-Azido-6-(Boc-amino)hexanoic acid (dicyclohexylammonium) salt, ≥97%
C11H20N4O4 · C12H23N