EMAIL THIS PAGE TO A FRIEND
Journal of Separation Science
Simultaneous separation and determination of phenolic acids, pentapeptides, and triterpenoid saponins in the root of Aster tataricus by high-performance liquid chromatography coupled with electrospray ionization quadrupole time-of-flight mass spectrometry.
PMID 25491750