EMAIL THIS PAGE TO A FRIEND
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association
Biological behavior of the intramucosal Helicobacter pylori-negative undifferentiated-type early gastric cancer: comparison with Helicobacter pylori-positive early gastric cancer.
PMID 25491775
Abstract
The differences in the growth morphology, proliferative ability, and background mucosa of the cancer between Helicobacter pylori (HP)-positive (HP+) gastric cancer (GC) and HP-negative (HP-) GC are still unclear. To clarify the differences, we compared the characteristics of the two types of cancer. Of the 91 patients with undifferentiated-type early GC who underwent endoscopic treatment at our hospital between August 2005 and April 2011, 23 HP- GC patients (all of whom had signet ring cell carcinoma measuring 20 mm or less in diameter) and 46 HP+ GC patients with signet ring cell carcinoma measuring 20 mm or less in diameter (out of a total of 68 HP+ GC patients) were enrolled in this study. Endoscopic atrophy and background mucosa were classified according to the updated Sydney system. The proliferative capacity of the cancer was assessed by examining the MIB-1 labeling index. With regard to the growth in the mucosal layer, the proportion of patients with cancer confined to the proliferative zone was significantly higher in the HP- GC group. Moderate or severer atrophy, intestinal metaplasia, mononuclear cell infiltration, and neutrophil infiltration according to the updated Sydney system were significantly commoner in the HP+ GC patients. Also, the MIB-1 labeling index was significantly higher in the HP+ GC group. HP+ GC appeared to show a higher proliferative capacity, more extensive spread, and more rapid progression, and inflammation associated with HP infection was suggested to be involved in the proliferation of this type of GC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU065891 MISSION® esiRNA, esiRNA human MKI67 (esiRNA1)